3-Amino-5-phenyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL393797-1GEA 37.5 EUR
SIAL393797-1G SIAL393797-5G
3-Amino-5-phenyl-1H-pyrazol, Sigma-Aldrich®
3-Amino-5-phenyl-1H-pyrazole
Formel: C₉H₉N₃
Molecular Weight: 159,19
MDL: MFCD00191749
CAS-Nummer: 1572-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR