4-Methyl-1H-pyrazol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
M1387-100MG M1387-1G M1387-500MG
SIALM1387-100MGEA 97.2 EUR
SIALM1387-100MG SIALM1387-1G SIALM1387-500MG
4-Methyl-1H-pyrazol Hydrochlorid, Sigma-Aldrich®
4-Methyl-1H-pyrazol Hydrochlorid
Formel: C₄H₇ClN₂
Molecular Weight: 118,5648 g/mol
MDL: MFCD00012707
CAS-Nummer: 56010-88-9