3-Amino-1H-pyrazol-4-carbonsäure
Lieferant: Thermo Scientific
MAYBGK02804.10EA 0 EUR
MAYBGK02804.10
3-Amino-1H-pyrazol-4-carbonsäure
3-Amino-1H-pyrazole-4-carboxylic acid
Formel: C₄H₅N₃O₂
Molecular Weight: 127,10 g/mol
MDL: MFCD00005239
CAS-Nummer: 41680-34-6
EINECS: 255-493-7