Bis(2-methoxyphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

SIAL710466-250MGEA 96.9 EUR
SIAL710466-250MG
Bis(2-methoxyphenyl)phosphin, Sigma-Aldrich®
Bis(2-methoxyphenyl)phosphin
Formel: C₁₄H₁₅O₂P
MDL: MFCD08064038
CAS-Nummer: 10177-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR