Dicyclopentylchlorphosphin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL649066-5GEA 260 EUR
SIAL649066-5G
Dicyclopentylchlorphosphin, Sigma-Aldrich®
Dicyclopentylchlorphosphin
Formel: C₁₀H₁₈ClP
MDL: MFCD06798131
CAS-Nummer: 130914-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR