Dicyclohexylchlorphosphin ≥98%

Lieferant: Strem Chemicals, Inc.
Gefahr

STRE15-1050-1GEA 86.7 EUR
STRE15-1050-1G
Dicyclohexylchlorphosphin ≥98%
Dicyclohexylchlorphosphin

Phosphine

Formel: C₁₂H₂₂ClP
Molecular Weight: 232,73 g/mol
Siedepunkt: 96…99 °C (0,1 mmHg)
Dichte: 1,054
MDL: MFCD00014294
CAS-Nummer: 16523-54-9
UN: 1760
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR