6-Methylchromon-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL417491-1GEA 35.4 EUR
SIAL417491-1G
6-Methylchromon-2-carbonsäure, Sigma-Aldrich®
6-Methylchromon-2-carbonsäure
Formel: C₁₁H₈O₄
MDL: MFCD00239435
CAS-Nummer: 5006-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR