6-Ethylchromon-2-carbonsäure

Lieferant: CHEMPUR

CHMPFL12038.POREA 0 EUR
FLUO012038.1G CHMPFL12038.POR FLUO012038.5G
6-Ethylchromon-2-carbonsäure
6-Ethylchromon-2-carbonsäure
Formel: C₁₂H₁₀O₄
MDL: MFCD03424502
CAS-Nummer: 5527-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR