6-Chlor-7-methylchromon

Lieferant: CHEMPUR

CHMPFL12050.POREA 0 EUR
CHMPFL12050.POR
6-Chlor-7-methylchromon
6-Chlor-7-methylchromon
Formel: C₁₀H₇ClO₂
Molecular Weight: 194,62
MDL: MFCD00239401
CAS-Nummer: 67029-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR