6-Fluorchromon-2-carbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL543764-1GEA
50
EUR
SIAL543764-1G
6-Fluorchromon-2-carbonsäure, Sigma-Aldrich®
6-Fluorchromon-2-carbonsäure
Formel:
C₁₀H₅FO₄ Molecular Weight: 208,15 g/mol |
MDL:
MFCD03070543 CAS-Nummer: 99199-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...