3-Formyl-6-nitrochromon

Lieferant: CHEMPUR

CHMPFL12014.POREA 0 EUR
CHMPFL12014.POR FLUO012014.5G
3-Formyl-6-nitrochromon
3-Formyl-6-nitrochromon
Formel: C₁₀H₅NO₅
Molecular Weight: 219,1504 g/mol
MDL: MFCD00192184
CAS-Nummer: 42059-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR