Chromon-3-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

SIAL383449-5GEA 34.8 EUR
SIAL383449-5G
Chromon-3-carbaldehyd, Sigma-Aldrich®
Chromon-3-carbaldehyd
Formel: C₁₀H₆O₃
Molecular Weight: 174,16 g/mol
MDL: MFCD00014667
CAS-Nummer: 17422-74-1
EINECS: 241-451-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR