3-Brom-6-fluorchromon

Lieferant: CHEMPUR

CHMPFL12062.POREA 0 EUR
CHMPFL12062.POR
3-Brom-6-fluorchromon
3-Brom-6-fluorchromon
Formel: C₉H₄BrFO₂
Molecular Weight: 243,03
MDL: MFCD03094003
CAS-Nummer: 179111-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR