3-Brom-6-chlorchromon

Lieferant: CHEMPUR

CHMPFL12057.POREA 0 EUR
CHMPFL12057.POR
3-Brom-6-chlorchromon
3-Brom-6-chlorchromon
Formel: C₉H₄BrClO₂
Molecular Weight: 259,49 g/mol
MDL: MFCD02093123
CAS-Nummer: 73220-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR