Tris(trimethylsilyl)methan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS21300.5EA 0 EUR
SILNSILS21300.5
Tris(trimethylsilyl)methan
Tris(trimethylsilyl)methan
Formel: C₁₀H₂₈Si₃
Molecular Weight: 232,59 g/mol
MDL: MFCD00008273
CAS-Nummer: 1068-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR