Tris(trimethylsilyl)methan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL302600-5GEA 586 EUR
SIAL302600-5G
Tris(trimethylsilyl)methan, Sigma-Aldrich®
Tris(trimethylsilyl)methan
Formel: C₁₀H₂₈Si₃
Molecular Weight: 232,59 g/mol
MDL: MFCD00008273
CAS-Nummer: 1068-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR