Dimethylethoxysilan
Lieferant: SILANE (PART OF CHEMPUR)
SILNSILS07250.25EA
121
EUR
SILNSILS07250.25
SILNSILS07250.100
SILNSILS07250.5
Dimethylethoxysilan
Dimethylethoxysilane
Formel:
C₄H₁₂OSi |
MDL:
MFCD00026750 CAS-Nummer: 14857-34-2 EINECS: 238-921-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...