N-(3-Fluorobenzyl)-3,5-dimethylaniline, 97%

H57055.03EA 153 EUR
ALFAH57055.03 ALFAH57055.MD
N-(3-Fluorobenzyl)-3,5-dimethylaniline, 97%
Purity: 97%
CAS No.: 1019591-65-1
Molecular Formula: C15H16FN
Formula Weight: 229.30
MDL No.: MFCD11145867
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR