N-(2-Fluorobenzyl)-2,4-dimethylaniline, 97%

H56861.MDEA 38.3 EUR
ALFAH56861.MD ALFAH56861.03
N-(2-Fluorobenzyl)-2,4-dimethylaniline, 97%
Purity: 97%
CAS No.: 1019618-03-1
Molecular Formula: C15H16FN
Formula Weight: 229.29
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR