2-(2-Thienyl)-4-thiazolcarbonsäure
Lieferant: Thermo Scientific
MAYBCC11501-0001EA
0
EUR
MAYBCC11501-0001
2-(2-Thienyl)-4-thiazolcarbonsäure
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formel:
C₈H₅NO₂S₂ Molecular Weight: 211,25 g/mol |
MDL:
MFCD01936014 CAS-Nummer: 24044-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...