2-(3-Pyridinyl)-4-thiazolcarbonsäure
Lieferant: Thermo Scientific
MAYBCC13701DAEA
0
EUR
MAYBCC13701DA
MAYBCC13701-0001
MAYBCC13701CB
2-(3-Pyridinyl)-4-thiazolcarbonsäure
2-(3-Pyridinyl)-4-thiazolcarbonsäure
Formel:
C₉H₆N₂O₂S Molecular Weight: 206,22 g/mol |
MDL:
MFCD00052304 CAS-Nummer: 39067-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...