2-Chlor-4-phenylthiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL688150-1GEA 45.4 EUR
SIAL688150-1G
2-Chlor-4-phenylthiazol, Sigma-Aldrich®
2-Chloro-4-phenylthiazol
Formel: C₉H₆ClNS
MDL: MFCD00956371
CAS-Nummer: 1826-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR