4-Chlor-2-(4-methoxyphenyl)chinazolin

Lieferant: CHEMPUR

CHMP79-0002.1EA 0 EUR
CHMP79-0002.1 CHMP79-0002.500
4-Chlor-2-(4-methoxyphenyl)chinazolin
4-Chloro-2-(4-methoxyphenyl)quinazoline
Formel: C₁₅H₁₁ClN₂O
MDL: MFCD03847393
CAS-Nummer: 55391-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR