4-Chlor-6-fluor-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0009.500EA 0 EUR
CHMP79-0009.500 CHMP79-0009.1
4-Chlor-6-fluor-2-phenylchinazolin
4-Chloro-6-fluoro-2-phenylquinazoline
Formel: C₁₄H₈ClFN₂
MDL: MFCD04115227
CAS-Nummer: 885277-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR