4-Chlor-2-(4-fluorphenyl)chinazolin

Lieferant: CHEMPUR

CHMP79-0003.500EA 0 EUR
CHMP79-0003.500 CHMP79-0003.1
4-Chlor-2-(4-fluorphenyl)chinazolin
4-Chloro-2-(4-fluorophenyl)quinazoline
Formel: C₁₄H₈ClFN₂
MDL: MFCD04115224
CAS-Nummer: 113242-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR