4-Chlor-2-(2-fluorphenyl)chinazolin

Lieferant: CHEMPUR

CHMP79-0004.500EA 0 EUR
CHMP79-0004.500 CHMP79-0004.1
4-Chlor-2-(2-fluorphenyl)chinazolin
4-Chloro-2-(2-fluorophenyl)quinazoline
Formel: C₁₄H₈ClFN₂
MDL: MFCD03030427
CAS-Nummer: 610276-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR