4-Chlor-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0001.1EA 0 EUR
CHMP79-0001.1 CHMP79-0001.500
4-Chlor-2-phenylchinazolin
4-Chloro-2-phenylquinazoline
Formel: C₁₄H₉ClN₂
MDL: MFCD00006713
CAS-Nummer: 6484-25-9
EINECS: 229-346-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR