4-Chlor-2-(4-chlorphenyl)chinazolin

Lieferant: CHEMPUR

CHMP79-0005.500EA 0 EUR
CHMP79-0005.500 CHMP79-0005.1
4-Chlor-2-(4-chlorphenyl)chinazolin
4-Chloro-2-(4-chlorophenyl)quinazoline
Formel: C₁₄H₈Cl₂N₂
MDL: MFCD03426664
CAS-Nummer: 59490-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR