4,6-Dichlor-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0007.500EA 0 EUR
CHMP79-0007.500 CHMP79-0007.1
4,6-Dichlor-2-phenylchinazolin
4,6-Dichloro-2-phenylquinazoline
Formel: C₁₄H₈Cl₂N₂
MDL: MFCD04115225
CAS-Nummer: 54665-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR