6-Brom-4-chlor-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0011.500EA 0 EUR
CHMP79-0011.500 CHMP79-0011.1
6-Brom-4-chlor-2-phenylchinazolin
6-Bromo-4-chloro-2-phenylquinazoline
Formel: C₁₄H₈BrClN₂
MDL: MFCD04115229
CAS-Nummer: 412923-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR