2-(4-Bromphenyl)-4-chlorchinazolin

Lieferant: CHEMPUR

CHMP79-0006.500EA 0 EUR
CHMP79-0006.500 CHMP79-0006.1
2-(4-Bromphenyl)-4-chlorchinazolin
2-(4-Bromophenyl)-4-chloroquinazoline
Formel: C₁₄H₈BrClN₂
MDL: MFCD01647442
CAS-Nummer: 83800-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR