4-Chlor-6-methyl-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0012.1EA 0 EUR
CHMP79-0012.1 CHMP79-0012.500
4-Chlor-6-methyl-2-phenylchinazolin
4-Chloro-6-methyl-2-phenylquinazoline
Formel: C₁₅H₁₁ClN₂
MDL: MFCD04115230
CAS-Nummer: 29083-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR