4-Chlor-5-methyl-2-phenylchinazolin

Lieferant: CHEMPUR

CHMP79-0013.500EA 0 EUR
CHMP79-0013.500 CHMP79-0013.1
4-Chlor-5-methyl-2-phenylchinazolin
4-Chloro-5-methyl-2-phenylquinazoline
Formel: C₁₅H₁₁ClN₂
MDL: MFCD04115231
CAS-Nummer: 885277-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR