2-(1,2-Benzisoxazol-3-yl)essigsäure
MAYBKM10833DAEA
0
EUR
MAYBKM10833DA
MAYBKM10833CB
2-(1,2-Benzisoxazol-3-yl)essigsäure
2-(1,2-Benzisoxazol-3-yl)essigsäure
Formel:
C₉H₇NO₃ Molecular Weight: 177,1568 g/mol |
MDL:
MFCD02180388 CAS-Nummer: 4865-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...