Bis(tetramethylcyclopentadienyl)chrom(II), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL562432-1GEA
99
EUR
SIAL562432-1G
Bis(tetramethylcyclopentadienyl)chrom(II), Sigma-Aldrich®
Bis(tetramethylcyclopentadienyl)chrom(II)
Formel:
C₁₈H₂₆Cr Molecular Weight: 294,40 g/mol |
MDL:
MFCD01862451 CAS-Nummer: 82066-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...