Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')eisen(III) ≥99% (99.9%-Fe)

Lieferant: Strem Chemicals, Inc.

STRE26-3910.5EA 0 EUR
STRE26-3910.5 ABCRAB120715.0025
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')eisen(III) ≥99% (99.9%-Fe)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')eisen(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₃₃H₅₇FeO₆
Molecular Weight: 605,65 g/mol
Siedepunkt: 300 °C (Zersetzung)
Schmelzpunkt: 164 °C
MDL: MFCD00067465
CAS-Nummer: 14876-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR