Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II) ≥98% (99.9%-Ni)

Lieferant: ABCR GMBH

ABCRAB123782.0001EA 0 EUR
ABCRAB123782.0001
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II) ≥98% (99.9%-Ni)
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₂₂H₃₈NiO₄
Molecular Weight: 425,23 g/mol
Siedepunkt: 90…110 °C (0,1 mmHg) (Subl.)
Schmelzpunkt: 223…225 °C
MDL: MFCD00192348
CAS-Nummer: 14481-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR