Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) ≥99%
Lieferant: Strem Chemicals, Inc.
STRE40-5000-25GEA
538
EUR
STRE40-5000-25G
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV) ≥99%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formel:
C₄₄H₇₆O₈Zr Molecular Weight: 824,29 g/mol Siedepunkt: 180 °C (0,1 mmHg) (Subl.) Schmelzpunkt: 318…320 °C |
MDL:
MFCD00145380 CAS-Nummer: 18865-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...