Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III) ≥98% (99.9%-Y)

Lieferant: Strem Chemicals, Inc.

STRE39-1000-1GEA 144 EUR
STRE39-1000-1G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III) ≥98% (99.9%-Y)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₃₃H₅₇O₆Y
Molecular Weight: 638,708 g/mol
Schmelzpunkt: 170…173 °C
MDL: MFCD00015713
CAS-Nummer: 15632-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR