Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)

Lieferant: ABCR GMBH

ABCRAB106364.0005EA 0 EUR
ABCRAB106364.0005
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Formel: C₃₃H₅₇O₆Y
Molecular Weight: 638,708 g/mol
MDL: MFCD00015713
CAS-Nummer: 15632-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR