Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)
Lieferant: ABCR GMBH
ABCRAB106202.0005EA
0
EUR
ABCRAB106202.0005
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)
Formel:
C₃₃H₅₇GdO₆ Molecular Weight: 707,052 g/mol |
MDL:
MFCD00010349 CAS-Nummer: 14768-15-1 EINECS: 238-834-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...