Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium Hydrat

Lieferant: Strem Chemicals, Inc.

STRE38-1000-25GEA 1750 EUR
STRE38-1000-25G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium Hydrat
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium Hydrat

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formel: C₂₂H₃₈O₄Sr·H₂O
Molecular Weight: 472,17 g/mol
Siedepunkt: 250 °C/ 230 °C (0,05 mmHg) (Zers./ Subl.)
Schmelzpunkt: 200…203 °C
MDL: MFCD00151017
CAS-Nummer: 199445-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR