Distilling heads
Lieferant: SciLabware
BIBBSH30/42/SCEA 2130.06 EUR
BIBBSH30/42/SC
Distilling heads
PYREX® borosilicate glass.
  • With integral jacketed condenser
  • Liquid leaving condenser is directed towards a GP Rotaflo® stopcock
  • Adjustment of stopcock controls proportion of liquid returned to the column or collected
  • Screwcap fitting for insertion of thermometer
  • Side-arm for venting or connection to vacuum supply
Fits on the top of a fractionating column to provide an effective liquid dividing head.
  • With integral jacketed condenser
  • Liquid leaving condenser is directed towards a GP Rotaflo® stopcock
  • Adjustment of stopcock controls proportion of liquid returned to the column or collected
  • Screwcap fitting for insertion of thermometer
  • Side-arm for venting or connection to vacuum supply