Disposable particle filtering respirators, Comfort-Line, zetMask®
Lieferant: ZVG
zetMask® comfort
ZVGA30953-01EA 0 EUR
ZVGA30953-01 ZVGA30941-01 ZVGA30943-01
Disposable particle filtering respirators, Comfort-Line, zetMask®
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
The particle-filtering half-masks of the Comfort line feature an extremely low breathing resistance combined with a high filtering capacity. This also applies for masks without exhalation valve.
  • High quality mask
  • Flat folding , individually hygienically packed
  • With elastic bands
  • Colour coded
Standards: EN 149:2001