Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Lieferant: Witeg
WITGDH.WOS501015EA 20 EUR
WITGDH.WOS501015 WITGDH.WOS501017 WITGDH.WOS501111 WITGDH.WOS501135 WITGDH.WOS501035 WITGDH.WOS501013 WITGDH.WOS501010 WITGDH.WOS501020 WITGDH.WOS501031
Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Rührer Rührwerkzeuge
Stirring impeller for high speed-type and high torque-type overhead stirrer.
  • Made of stainless steel 304 (high-grade)
  • Resistant to acids, chemicals, corrosion and high temperature