Overhead stirrers, HT-50AX, Wisd
Lieferant: Witeg
WITGDH.WOS01251EA 461.42 EUR
WITGDH.WOS01251 WITGDH.WOS01252
Overhead stirrers, HT-50AX, Wisd
Rührer Rührwerke
These overhead stirrers are designed for chemical, biological, medical or pharmaceutical applications. The device is especially suited for use with low viscosity liquids which become highly viscous during the stirring process.
  • Long life due to simple and solid construction
  • Constant speed in spite of viscosity fluctuation
  • Motor features overheat protection
  • Smooth increase of speed for improved safety
Stepless variable adjustability of every stirrer concerning to the continuous bored chuck.
Lieferumfang: Includes Allen key, body holder, chuck key, operating manual. In addition a stand, clamp and stirring tool are included depending on model.