Multi-position magnetic stirrers, WiseStir® MS-MP series
Lieferant: Witeg
WiseStir®
WITGDH.WMS03038EA 482.67 EUR
WITGDH.WMS03038 WITGDH.WMS03036
Multi-position magnetic stirrers, WiseStir® MS-MP series
Rührer Mehrstellen-Magnetrührer
Stainless steel. This is a digital multi-position magnetic stirrer without heating, controlled by microprocessor. Modules consist of 4 or 8 stirring positions on a stainless steel platform, with a single electrical connection. Ideally suited for use in incubators and climatic chambers.
  • Digital LCD display for RPM and status
  • Locking mode supported for experimental safety
  • Smooth start and change of the stirring speed