Pulvertrichter
Lieferant: Witeg
WITG5.790.004EA 17.81 EUR
WITG5.790.004 WITG5.790.001
Pulvertrichter
Trichter Pulvertrichter
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes