Desiccators, NOVUS
Lieferant: Witeg
WITG5.660.200EA 320.66 EUR
WITG5.660.200
Desiccators, NOVUS
Exsikkatoren Exsikkatoren, Glas
This desiccator is made from DURAN® glass, without porcelain disc, with tubulated lid.