Gewebehomogenisatoren nach Potter-Elvehjem
Lieferant: Witeg
WITG5.553.734EA 93.81 EUR
WITG5.553.734 WITG5.553.735 WITG5.553.737 WITG5.553.738
Gewebehomogenisatoren nach Potter-Elvehjem
Homogenisatoren Gewebehomogenisatoren
Tissue grinders are made of borosilicate glass, with PTFE pestle and stainless steel shaft.
Clearance between pestle and tube is 102 to 152 μm.